Få mere at vide
1-(4-Methylbenzyl)-1,4,8,11-tetraazacyclotetradecane (>85%), TRC
Artikelnummer. 30469511
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469511 500 mg
30468411 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30469511 Leverandør Toronto Research Chemicals Leverandørnr. TRCM312280500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 351472-73-6
Molekylær formel C18H32N4
Molekylvægt (g/mol) 304.47
Synonym Plerixafor Impurity J
SMIL Cc1ccc(CN2CCCNCCNCCCNCC2)cc1
Tekniske data
Kemisk navn eller materiale 1-(4-Methylbenzyl)-1,4,8,11-tetraazacyclotetradecane
Mængde 500 mg
InChI formel InChI=1S/C18H32N4/c1-17-4-6-18(7-5-17)16-22-14-3-10-20-12-11-19-8-2-9-21-13-15-22/h4-7,19-21H,2-3,8-16H2,1H3