Få mere at vide
1-(4’-Methoxyphenyl)proanol-methyl-d3, TRC
Artikelnummer. 30468063
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30468063 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468063 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26622750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(4'-Methoxyphenyl)proanol-methyl-d3
CAS 1189968-85-1
Molekylær formel C10H11D3O2
Mængde 50 mg
Synonym alpha-(Ethyl-3,3,3-d3)-4-methoxy-benzenemethanol, alpha-Ethyl-p-methoxy-benzyl Alcohol-d3, NSC 1264-d3, NSC 406923-d3
InChI formel InChI=1S/C10H14O2/c1-3-10(11)8-4-6-9(12-2)7-5-8/h4-7,10-11H,3H2,1-2H3/i1D3
SMIL [2H]C([2H])([2H])CC(O)c1ccc(OC)cc1
IUPAC navn 3,3,3-trideuterio-1-(4-methoxyphenyl)propan-1-ol
Molekylvægt (g/mol) 169.24
Formel vægt 169.1182