missing translation for 'onlineSavingsMsg'
Få mere at vide

1-(4-Methoxyphenyl)acetophenon, TRC

Artikelnummer. 30472111
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30472111 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472111 Supplier Toronto Research Chemicals Supplier No. TRCM2658005MG
 Begrænset vare
Forventet forsendelse: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1-(4-Methoxyphenyl)acetophenone
CAS 1023-17-2
Molekylær formel C15H14O2
Mængde 5 mg
Synonym 1-(4-Methoxyphenyl)-2-phenyl-ethanone, 2-Phenyl-p-methoxyacetophenone, 4-Methoxy-alpha-phenylacetophenone, 4-Methoxydeoxybenzoin, Benzyl p-Methoxyphenyl Ketone, NSC 26658, p-Anisyl Benzyl Ketone
InChI formel InChI=1S/C15H14O2/c1-17-14-9-7-13(8-10-14)15(16)11-12-5-3-2-4-6-12/h2-10H,11H2,1H3
SMIL COc1ccc(cc1)C(=O)Cc2ccccc2
IUPAC navn 1-(4-methoxyphenyl)-2-phenylethanon
Molekylvægt (g/mol) 226.27
Formel vægt 226.0994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.