Få mere at vide
1-(4-Hydroxy-3-methoxyphenyl)propan-1-one, Mikromol™
Artikelnummer. 30051903
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051903 25 mg
30050912 100 mg
30464138 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051903 Leverandør Mikromol Leverandørnr. MM3659.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(4-Hydroxy-3-methoxyphenyl)propan-1-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(4-Hydroxy-3-methoxyphenyl)propan-1-one
CAS 1835-14-9
Analyt- eller komponentnavne 1-(4-Hydroxy-3-methoxyphenyl)propan-1-one
Forsendelsestilstand Room Temperature
Molekylær formel C10 H12 O3
Certificeringer/compliance ISO 17025
Synonym 1-Propanone, 1-(4-hydroxy-3-methoxyphenyl)-, Propiophenone, 4'-hydroxy-3'-methoxy- (6CI,7CI,8CI), 1-(3-Methoxy-4-hydroxyphenyl)-1-propanone, 1-(4-Hydroxy-3-methoxyphenyl)-1-propanone, 1-(4-Hydroxy-3-methoxyphenyl)propan-1-one, 1-(4-Hydroxy-3-methoxyphenyl)propanone, 1-Propiovanillone, 4'-Hydroxy-3'-methoxypropiophenone, Ethyl guaiacyl ketone, Guaiacyl-1-propanone, NSC 16691, Propioguaiacol, Propioguaiacone, Propiovanillone, α-Guaiacyl ethyl ketone, α-Guaiacylpropanone
InChI formel InChI=1S/C10H12O3/c1-3-8(11)7-4-5-9(12)10(6-7)13-2/h4-6,12H,3H2,1-2H3
SMIL CCC(=O)c1ccc(O)c(OC)c1
IUPAC navn 1-(4-hydroxy-3-methoxyphenyl)propan-1-one
Molekylvægt (g/mol) 180.2
Mængde 25 mg
Formel vægt 180.0786 g/mol