missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluoro-2-nitrophenyl)homopiperazine, 97%, Thermo Scientific™

Catalog Number 15480739
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
874814-95-6
C11H14FN3O2
239.25
HFLSWHSIIVSRPS-UHFFFAOYSA-N
1-4-fluoro-2-nitrophenyl-1,4-diazepane, 1-4-fluoro-2-nitrophenyl-1,4-diazaperhydroepine, 1h-1,4-diazepine, 1-4-fluoro-2-nitrophenyl hexahydro
18801299
1-(4-fluoro-2-nitrophenyl)-1,4-diazepane
C1CNCCN(C1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
This item is not returnable. View return policy
874814-95-6
C11H14FN3O2
239.25
HFLSWHSIIVSRPS-UHFFFAOYSA-N
1-4-fluoro-2-nitrophenyl-1,4-diazepane, 1-4-fluoro-2-nitrophenyl-1,4-diazaperhydroepine, 1h-1,4-diazepine, 1-4-fluoro-2-nitrophenyl hexahydro
18801299
1-(4-fluoro-2-nitrophenyl)-1,4-diazepane
C1CNCCN(C1)C2=C(C=C(C=C2)F)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3259
Formula Weight 239.24
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)homopiperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'