missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluoro-2-nitrophenyl)azepane, 97%

Brand:  Alfa Aesar™ H63210.06

Additional Details : Weight : 0.00500kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15479778

  • 2132.98 DKK / 5g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

897598-47-9
1-(4-Fluoro-2-nitrophenyl)hexamethyleneimine

Specifications

1-(4-Fluoro-2-nitrophenyl)azepane
897598-47-9
1-(4-Fluoro-2-nitrophenyl)hexamethyleneimine
Solid
5g
UN3259
238.25
97%
Videos
SDS Documents