Få mere at vide
(±)-1,4-Dithiothreitol-d10, CDN
Artikelnummer. 30319074
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319074 0.05 g -
30318527 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319074 Leverandør CDN Leverandørnr. CDND39310.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Dithiothreitol-D10
CAS 203633-21-0
Molekylær formel C4 D10 O2 S2
Mængde 0.05 g
Synonym 2,3-Butane-1,1,2,3,4,4-d6-diol-d2, 1,4-di(mercapto-d)-, (2 R,3 R)-rel- (9 CI), 2,3-Butane-1,1,2,3,4,4-d6-diol-d2, 1,4-di(mercapto-d)-, (R*,R*)- (ZCI), (2 R,3 R)-rel-1,4-Di(mercapto-d)-2,3-butane-1,1,2,3,4,4-d6-diol-d2, (±)-1,4-Dithiothreitol-d10, DL-Dithiothreitol-D10, DTT-d10
InChI formel InChI=1 S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8 H,1-2H2/t3-,4-/m1/s1/i1D2,2D2,3 D,4 D,5 D,6 D/hD2
SMIL [2 H]O[C@]([2 H])(C([2 H])([2 H])S[2 H])[C@]([2 H])(O[2 H])C([2 H])([2 H])S[2 H]
IUPAC navn rac-(2 R,3 R)-1,1,2,3,4,4-hexadeuterio-2,3-dideuteriooxy-1,4-bis(deuteriosulfanyl)butane
Molekylvægt (g/mol) 164.3126
Formel vægt 164.075 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity