Få mere at vide
(±)-1,4-Dithiothreitol-1,1,2,3,4,4-d6, CDN
Artikelnummer. 30310233
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30310233 0.1 g 0.1g
30309685 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310233 Leverandør CDN Leverandørnr. CDND66310.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Dithiothreitol-1,1,2,3,4,4-D6
CAS 850153-85-4
Molekylær formel C4 D6 H4 O2 S2
Mængde 0.1 g
Synonym 2,3-Butane-1,1,2,3,4,4-d6-diol, 1,4-dimercapto-, (2 R,3 R)-rel- (9 CI), (2 R,3 R)-rel-1,4-Dimercapto-2,3-butane-1,1,2,3,4,4-d6-diol, (±)-1,4-Dithiothreitol-1,1,2,3,4,4-d6, DL-Dithiothreitol-1,1,2,3,4,4-D6, DTT-d6
InChI formel InChI=1 S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8 H,1-2H2/t3-,4-/m1/s1/i1D2,2D2,3 D,4 D
SMIL [2 H]C([2 H])[C@@]([2 H])(O)[C@]([2 H])(O)C([2 H])([2 H])S
IUPAC navn rac-(2 R,3 R)-1,1,2,3,4,4-hexadeuterio-1,4-bis(sulfanyl)butane-2,3-diol
Molekylvægt (g/mol) 160.288
Formel vægt 160.0499 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity