Få mere at vide
1,4-Dioxane D8 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30315210
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30315210 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315210 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA12865010AL1000
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Dioxane D8
CAS 17647-74-4
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C4 2H8 O2
Mængde 1 mL
Synonym p-Dioxane-d8 (8CI), 1,4-Dioxane-d8, Octadeutero-p-dioxane, Octadeuterodioxane, Perdeuterio p-dioxane, p-Dioxane-2,2,3,3,5,5,6,6-d8
InChI formel InChI=1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2/i1D2,2D2,3D2,4D2
SMIL [2H]C1([2H])OC([2H])([2H])C([2H])([2H])OC1([2H])[2H]
IUPAC navn 2,2,3,3,5,5,6,6-octadeuterio-1,4-dioxane
Molekylvægt (g/mol) 96.15
Formel vægt 96.1026 g/mol