Få mere at vide
1,4-Dioxacyclotetradecane-5,14-dione, TRC
Artikelnummer. 17958504
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17958504 10 mg
17938984 5 mg
17927264 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17958504 Leverandør Toronto Research Chemicals Leverandørnr. TRCD77554010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5578-82-5
Molekylær formel C12 H20 O4
Molekylvægt (g/mol) 228.28
Synonym Cyclic Ethylene Ester Sebacic Acid, Cyclic Ethylene Decanedioate, Cyclic 1,2-Ethanediyl Ester Decanedioic Acid, Cyclic Sebacate Ethylene Glycol, Ethylene Sebacate;
IUPAC navn 1,4-dioxacyclotetradecane-5,14-dione
SMIL O=C1CCCCCCCCC(=O)OCCO1
Tekniske data
Kemisk navn eller materiale 1,4-Dioxacyclotetradecane-5,14-dione
Mængde 10 mg
InChI formel InChI=1S/C12H20O4/c13-11-7-5-3-1-2-4-6-8-12(14)16-10-9-15-11/h1-10H2
Formel vægt 228.1362