missing translation for 'onlineSavingsMsg'
Få mere at vide

1,4-Dioxacyclododecane-5,12-dione, TRC

Artikelnummer. 30369751
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30369751 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30369751 Leverandør Toronto Research Chemicals Leverandørnr. TRCD48932310MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1,4-Dioxacyclododecane-5,12-dione
CAS 90073-95-3
Molekylær formel C10 H16 O4
Mængde 10 mg
InChI formel InChI=1S/C10H16O4/c11-9-5-3-1-2-4-6-10(12)14-8-7-13-9/h1-8H2
SMIL O=C1CCCCCCC(=O)OCCO1
IUPAC navn 1,4-dioxacyclododecane-5,12-dione
Molekylvægt (g/mol) 200.232
Formel vægt 200.105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.