Få mere at vide
1,4-Dimethoxybenzene-d10, CDN
Artikelnummer. 30319813
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319813 0.1 g
30317437 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319813 Leverandør CDN Leverandørnr. CDND51770.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Dimethoxybenzene D10
CAS 74079-00-8
Molekylær formel C8 D10 O2
Mængde 0.1 g
Synonym Benzene-1,2,4,5-d4, 3,6-di(methoxy-d3)- (9 CI), p-Dimethoxybenzene-d10, 1,4-Di(methoxy-d3)benzene-2,3,5,6-d4, 1,4-Bis(trideuteriomethoxy)-2,3,5,6-tetradeuteriobenzene, 4-Methoxyanisole-d10, Quinol dimethyl ether-d10, p-Hydroquinone dimethyl ether-d10, p-Methoxyanisole-d10, 1,4-Dimethoxybenzene-d10, 1,4-Dimethoxybenzene-D10
InChI formel InChI=1 S/C8H10O2/c1-9-7-3-5-8(10-2)6-4-7/h3-6 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c(OC([2 H])([2 H])[2 H])c([2 H])c([2 H])c1OC([2 H])([2 H])[2 H]
IUPAC navn 1,2,4,5-tetradeuterio-3,6-bis(trideuteriomethoxy)benzene
Molekylvægt (g/mol) 148.225
Formel vægt 148.131 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity