Få mere at vide
1,4-Dihydro-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester, TRC
Artikelnummer. 30377844
Skift visning
Mængde:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377844 25 g
30368796 5 g
30378610 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30377844 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44974025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52980-28-6
Molekylær formel C12H11NO3
Molekylvægt (g/mol) 217.22
Synonym 3-Ethoxycarbonyl-4(1H)-quinolone, 4-Oxo-1,4-dihydroquinoline-3-carboxylic acid ethyl ester, Ethyl 1,4-dihydro-4-oxoquinoline-3-carboxylate, Ethyl 4-oxo-1,4-dihydroquinoline-3-carboxylate
SMIL CCOC(=O)C1=CNc2ccccc2C1=O
Tekniske data
Kemisk navn eller materiale 1,4-Dihydro-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester
Mængde 25 g
InChI formel InChI=1S/C12H11NO3/c1-2-16-12(15)9-7-13-10-6-4-3-5-8(10)11(9)14/h3-7H,2H2,1H3,(H,13,14)
Formel vægt 217.0739