Få mere at vide
1,4-Dihydro-1-(5-methoxy-1H-benzimidazol-2-yl)-3,5-dimethyl-4-oxo-2-pyridinecarboxylic Acid Ammonium Salt (>85%), TRC
Artikelnummer. 30137650
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137650 10 mg
30010271 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137650 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44959510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2227107-89-1
Molekylær formel C16 H14 N3 O4 . H4 N
Molekylvægt (g/mol) 330.34
Synonym 2-Pyridinecarboxylic acid, 1,4-dihydro-1-(6-methoxy-1H-benzimidazol-2-yl)-3,5-dimethyl-4-oxo-, ammonium salt (1:1), 1,4-Dihydro-1-(6-methoxy-1H-benzimidazol-2-yl)-3,5-dimethyl-4-oxo-2-pyridinecarboxylic acid ammonium salt, 1-(5-Methoxy-1H-benzoimidazol-2-yl)-3,5-dimethyl-4-oxo-1,4-dihydropyridine-2-carboxylic acid ammonium salt, 1,4-Dihydro-1-(5-methoxy-1H-benzimidazol-2-yl)-3,5-dimethyl-4-oxo-2-pyridinecarboxylic acid ammonium salt, 1-(5-Methoxy-1H-benzoimidazol-2-yl)-3,5-dimethyl-4-oxo-1,4-dihydropyridine-2-carboxylic Acid Ammonium Salt
IUPAC navn azanium;1-(5-methoxy-1H-benzimidazol-2-yl)-3,5-dimethyl-4-oxopyridine-2-carboxylate
SMIL [NH4+].COc1ccc2[nH]c(nc2c1)N3C=C(C)C(=O)C(=C3C(=O)[O-])C
Tekniske data
Kemisk navn eller materiale 1,4-Dihydro-1-(5-methoxy-1H-benzimidazol-2-yl)-3,5-dimethyl-4-oxo-2-pyridinecarboxylic Acid Ammonium Salt
Mængde 10 mg
InChI formel InChI=1S/C16H15N3O4.H3N/c1-8-7-19(13(15(21)22)9(2)14(8)20)16-17-11-5-4-10(23-3)6-12(11)18-16;/h4-7H,1-3H3,(H,17,18)(H,21,22);1H3
Formel vægt 330.1328