Få mere at vide
1,4-Diethylbenzene-d14, CDN
Artikelnummer. 30308900
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30308900 0.25 g -
30316695 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308900 Leverandør CDN Leverandørnr. CDND63100.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Diethylbenzene-D14
CAS 1219803-64-1
Molekylær formel C10 D14
Mængde 0.25 g
Synonym Benzene-1,2,4,5-d4, 3,6-di(ethyl-1,1,2,2,2-d5)- (ACI), 3,6-Di(ethyl-1,1,2,2,2-d5)benzene-1,2,4,5-d4 (ACI), 1,4-Di(ethyl-1,1,2,2,2-d5)benzene-2,3,5,6-d4, Perdeutero-1,4-diethylbenzene, 1,4-Diethylbenzene-d14, 1,4-Diethylbenzene-D14
InChI formel InChI=1 S/C10H14/c1-3-9-5-7-10(4-2)8-6-9/h5-8 H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2,5 D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,2,4,5-tetradeuterio-3,6-bis(1,1,2,2,2-pentadeuterioethyl)benzene
Molekylvægt (g/mol) 148.3044
Formel vægt 148.1974 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity