Få mere at vide
1,4-Dichlorobutane-d8, TRC
Artikelnummer. 30370505
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30370505 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370505 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4360095MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Dichlorobutane-d8
CAS 83547-96-0
Molekylær formel C4 D8 Cl2
Mængde 5 mg
Synonym Butane-1,1,2,2,3,3,4,4-d8, 1,4-dichloro- (9CI), 1,4-Dichlorobutane-d8, 1,4-Dichlorobutane-1,1,2,2,3,3,4,4-d8, 1,4-Dichlorobutane-D8, 4-Chlorobutyl-d8 chloride
InChI formel InChI=1S/C4H8Cl2/c5-3-1-2-4-6/h1-4H2/i1D2,2D2,3D2,4D2
SMIL [2H]C([2H])(Cl)C([2H])([2H])C([2H])([2H])C([2H])([2H])Cl
IUPAC navn 1,4-dichloro-1,1,2,2,3,3,4,4-octadeuteriobutane
Molekylvægt (g/mol) 135.062
Formel vægt 134.051