Få mere at vide
1,4-Dichlorobenzene-d4, CDN
Artikelnummer. 30310810
Change view
Click to view available options
Mængde:
5 g
Product Code. Mængde
30310810 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30310810 Supplier CDN Supplier No. CDND10345G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Kemisk navn eller materiale 1,4-Dichlorobenzene-D4
CAS 3855-82-1
Molekylær formel C6 D4 Cl2
Mængde 5 g
Synonym 1,4-Dichlorobenzene D4, Benzene-1,2,4,5-d4, 3,6-dichloro-, Benzene-1,2,4,5-d4, dichloro- (6 CI,7 CI), 3,6-Dichlorobenzene-1,2,4,5-d4, 1,4-Dichlorobenzene-d4, 1,4-Dichloroperdeuteriobenzene, Tetradeutero-p-dichlorobenzene, p-Dichlorobenzene-d4
InChI formel InChI=1 S/C6H4Cl2/c7-5-1-2-6(8)4-3-5/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(Cl)c([2 H])c([2 H])c1Cl
IUPAC navn 1,4-dichloro-2,3,5,6-tetradeuteriobenzene
Molekylvægt (g/mol) 151.027
Formel vægt 149.994 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity