missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-2-butyne, 99%, Thermo Scientific Chemicals

Catalog Number p-103957
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
821-10-3
C4H4Cl2
122.98
MFCD00000981
RCHDLEVSZBOHOS-UHFFFAOYSA-N
1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference
13182
1,4-dichlorobut-2-yne
ClCC#CCCl
This item is not returnable. View return policy
821-10-3
C4H4Cl2
122.98
MFCD00000981
RCHDLEVSZBOHOS-UHFFFAOYSA-N
1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference
13182
1,4-dichlorobut-2-yne
ClCC#CCCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.2500g/mL
Boiling Point 165°C to 168°C
Flash Point 160°C
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClCH2C∽CCH2Cl
Refractive Index 1.5030 to 1.506
Quantity 25 mL
Specific Gravity 1.25
Formula Weight 122.98
Percent Purity 99%
Chemical Name or Material 1, 4-Dichloro-2-butyne, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,4-Dichloro-2-butyne
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'