Få mere at vide
1,4-Dibromobutane-2,2,3,3-d4, CDN
Artikelnummer. 30326620
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326620 5 g
30311845 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326620 Leverandør CDN Leverandørnr. CDND12325G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Dibromobutane-2,2,3,3-D4
CAS 52089-63-1
Molekylær formel C4 D4 H4 Br2
Mængde 5 g
Synonym Butane-2,2,3,3-d4, 1,4-dibromo- (7 CI, 9 CI, ACI), 1,4-Dibromobutane-2,2,3,3-d4 (ACI), 1,4-Dibromobutane-d4, 1,4-Dibromobutane-2,2,3,3-D4
InChI formel InChI=1 S/C4H8Br2/c5-3-1-2-4-6/h1-4H2/i1D2,2D2
SMIL [2 H]C([2 H])(CBr)C([2 H])([2 H])CBr
IUPAC navn 1,4-dibromo-2,2,3,3-tetradeuteriobutane
Molekylvægt (g/mol) 219.939
Formel vægt 217.9244 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity