Få mere at vide
1,4-Diazabicyclo[2.2.2]octane-d12, CDN
Artikelnummer. 30320780
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320780 0.1 g 0.1g
30318823 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320780 Leverandør CDN Leverandørnr. CDND68580.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Diazabicyclo[2.2.2 ]octane-D12
CAS 119451-78-4
Molekylær formel C6 D12 N2
Mængde 0.1 g
Synonym 1,4-Diazabicyclo[2.2.2 ]octane-2,2,3,3,5,5,6,6,7,7,8,8-d12 (9 CI), 1,4-Diazabicyclo[2.2.2 ]octane-d12, Dabco-d12, 1,4-Diazabicyclo[2.2.2 ]octane-D12
InChI formel InChI=1 S/C6H12N2/c1-2-8-5-3-7(1)4-6-8/h1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2
SMIL [2 H]C1([2 H])N2C([2 H])([2 H])C([2 H])([2 H])N(C([2 H])([2 H])C2([2 H])[2 H])C1([2 H])[2 H]
IUPAC navn 2,2,3,3,5,5,6,6,7,7,8,8-dodecadeuterio-1,4-diazabicyclo[2.2.2 ]octane
Molekylvægt (g/mol) 124.2468
Formel vægt 124.1754 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity