Få mere at vide
1,4-Di(hydroxymethyl)benzene-d4, TRC
Artikelnummer. 30375479
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30375479 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375479 Leverandør Toronto Research Chemicals Leverandørnr. TRCD452347100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Di(hydroxymethyl)benzene-d4
CAS 1158734-28-1
Molekylær formel C8 D4 H6 O2
Mængde 100 mg
Synonym 1,4-Benzene-2,3,5,6-d4-dimethanol, 1,4-Di(hydroxymethyl)benzene-d4, [2,3,5,6-Tetradeuterio-4-(hydroxymethyl)phenyl]methanol, 1,4-Di(hydroxymethyl)benzene-2,3,5,6-D4, 1,4-Phenylenedimethanol-d4, 1,4-Xylylene glycol-d4, α,α'-Dihydroxy-p-xylene-d4
InChI formel InChI=1S/C8H10O2/c9-5-7-1-2-8(6-10)4-3-7/h1-4,9-10H,5-6H2/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(CO)c([2H])c([2H])c1CO
IUPAC navn [2,3,5,6-tetradeuterio-4-(hydroxymethyl)phenyl]methanol
Molekylvægt (g/mol) 142.188
Formel vægt 142.093