Få mere at vide
1,4-Di(bromomethyl)benzene-d4, TRC
Artikelnummer. 30370997
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30370997 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370997 Leverandør Toronto Research Chemicals Leverandørnr. TRCD42565250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Di(bromomethyl)benzene-d4
CAS 74903-75-6
Molekylær formel C8H4D4Br2
Mængde 50 mg
Synonym 1,4-Bis(bromomethyl)benzene-d4, 1,4-Xylylene-d4 Dibromide, 4-Bromomethylbenzyl-d4 Bromide, p-Xylylene-d4 Bromide, NSC 6226-d4
InChI formel InChI=1S/C8H8Br2/c9-5-7-1-2-8(6-10)4-3-7/h1-4H,5-6H2/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(CBr)c([2H])c([2H])c1CBr
IUPAC navn 1,4-bis(bromomethyl)-2,3,5,6-tetradeuteriobenzene
Molekylvægt (g/mol) 267.98
Formel vægt 265.9244