missing translation for 'onlineSavingsMsg'
Få mere at vide

1,4-Cyclohexanedione-d8, TRC

Artikelnummer. 30358226
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30358226 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30358226 Supplier Toronto Research Chemicals Supplier No. TRCC98799210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1,4-Cyclohexanedione-d8
CAS 23034-25-5
Molekylær formel C6D8O2
Mængde 10 mg
Synonym 1,4-Cyclohexanedione-2,2,3,3,5,5,6,6-d8
InChI formel InChI=1S/C6H8O2/c7-5-1-2-6(8)4-3-5/h1-4H2/i1D2,2D2,3D2,4D2
SMIL O=C(C([2H])([2H])C1([2H])[2H])C([2H])([2H])C([2H])([2H])C1=O
IUPAC navn cyclohexane-1,4-dione-d8
Molekylvægt (g/mol) 120.18
Formel vægt 120.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.