Få mere at vide
1,4-Cyclohexanedione-d8, TRC
Artikelnummer. 30351737
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30351737 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30351737 Leverandør Toronto Research Chemicals Leverandørnr. TRCC987992100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Cyclohexanedione-d8
CAS 23034-25-5
Molekylær formel C6D8O2
Mængde 100 mg
Synonym 1,4-Cyclohexanedione-2,2,3,3,5,5,6,6-d8
InChI formel InChI=1S/C6H8O2/c7-5-1-2-6(8)4-3-5/h1-4H2/i1D2,2D2,3D2,4D2
SMIL O=C(C([2H])([2H])C1([2H])[2H])C([2H])([2H])C([2H])([2H])C1=O
IUPAC navn cyclohexane-1,4-dione-d8
Molekylvægt (g/mol) 120.18
Formel vægt 120.1