missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,4-Cyclohexanedione bis(ethylene acetal), 99%

Brand:  Alfa Aesar™ B20784.18

Additional Details : CAS Number : 183-97-1 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11495676

  • 1234.00 DKK / 50g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

183-97-1
200.234
YSMVSEYPOBXSOK-UHFFFAOYSA-N
135986
C1CC2(CCC13OCCO3)OCCO2
C10H16O4
MFCD00010851
1,4-cyclohexanedione bis ethylene ketal, 1,4,9,12-tetraoxadispiro 4.2.4.2 tetradecane, 1,4-cyclohexanedione bis ethylene acetal, 1,4,9,12-tetraoxadispiro 4,2,4,2 tetradecane, 1,4-cyclohexanedione bis ethyleneketal, 1,4,9,12-tetraoxadispiro 4.2.4^ 8 .2^ 5 tetradecane, 1,4,9,12-tetraoxadispiro 4.2.4?.2? tetradecane, maybridge1_006651, 1,4-cyclohexanedione ethyleneketal
1,4,9,12-tetraoxadispiro[4.2.4^{8}.2^{5}]tetradecane

Specifications

1,4-Cyclohexanedione bis(ethylene acetal)
79°C to 80°C
183-97-1
MFCD00010851
YSMVSEYPOBXSOK-UHFFFAOYSA-N
1,4,9,12-tetraoxadispiro[4.2.4^{8}.2^{5}]tetradecane
135986
99%
75°C to 85°C (0.5mmHg)
50g
C10H16O4
1,4-cyclohexanedione bis ethylene ketal, 1,4,9,12-tetraoxadispiro 4.2.4.2 tetradecane, 1,4-cyclohexanedione bis ethylene acetal, 1,4,9,12-tetraoxadispiro 4,2,4,2 tetradecane, 1,4-cyclohexanedione bis ethyleneketal, 1,4,9,12-tetraoxadispiro 4.2.4^ 8 .2^ 5 tetradecane, 1,4,9,12-tetraoxadispiro 4.2.4?.2? tetradecane, maybridge1_006651, 1,4-cyclohexanedione ethyleneketal
C1CC2(CCC13OCCO3)OCCO2
200.234
200.23
Videos
SDS Documents