Få mere at vide
1-(4-Chlorophenyl)cyclobutane-d6 Carbonitrile, TRC
Artikelnummer. 30367920
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30367920 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367920 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37951210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(4-Chlorophenyl)cyclobutane-d6 Carbonitrile
CAS 1217114-23-2
Molekylær formel C11H4D6ClN
Mængde 10 mg
Synonym 1-(4-Chlorophenyl)cyclobutanecarbonitrile-d6, 1-(p-Chlorophenyl)cyclobutanecarbonitrile-d6, NSC 154613-d6
InChI formel InChI=1S/C11H10ClN/c12-10-4-2-9(3-5-10)11(8-13)6-1-7-11/h2-5H,1,6-7H2/i1D2,6D2,7D2
SMIL [2H]C1([2H])C([2H])([2H])C(C#N)(c2ccc(Cl)cc2)C1([2H])[2H]
IUPAC navn 1-(4-chlorophenyl)-2,2,3,3,4,4-hexadeuteriocyclobutane-1-carbonitrile
Molekylvægt (g/mol) 197.69