Få mere at vide
1-(4-Chlorophenyl)-5-[6-[[4-[(4-chlorophenyl)amino]-6-[(1S,2R,3R,4R)-1,2,3,4,5-pentahydroxypentyl]-1,3,5-triazin-2-yl]amino]hexyl]biguanide D-Gluconic Acid, TRC
Artikelnummer. 30134691
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134691 500 mg
30012623 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134691 Leverandør Toronto Research Chemicals Leverandørnr. TRCC378545500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3046108-31-7
Molekylær formel C28H38Cl2N10O5 • C6H12O7
Molekylvægt (g/mol) 861.73
Synonym Chlorhexidine Impurity J D-Gluconic Acid;
IUPAC navn 1-[amino-(4-chloroanilino)methylidene]-2-[6-[[4-(4-chloroanilino)-6-[(1S,2R,3R,4R)-1,2,3,4,5-pentahydroxypentyl]-1,3,5-triazin-2-yl]amino]hexyl]guanidine;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
SMIL N=C(NC(NCCCCCCNC1=NC(NC2=CC=C(Cl)C=C2)=NC([C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO)=N1)=N)NC3=CC=C(Cl)C=C3.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O
Tekniske data
Kemisk navn eller materiale 1-(4-Chlorophenyl)-5-[6-[(4-[(4-chlorophenyl)amino]-6-[(1S,2R,3R,4R)-1,2,3,4,5-pentahydroxypentyl]-1,3,5-triazin-2-yl)amino]hexyl]biguanide D-Gluconic Acid Salt
Mængde 500 mg
InChI formel InChI=1S/C28H38Cl2N10O5.C6H12O7/c29-16-5-9-18(10-6-16)35-26(32)39-25(31)33-13-3-1-2-4-14-34-27-37-24(23(45)22(44)21(43)20(42)15-41)38-28(40-27)36-19-11-7-17(30)8-12-19;7-1-2(8)3(9)4(10)5(11)6(12)13/h5-12,20-23,41-45H,1-4,13-15H2,(H5,31,32,33,35,39)(H2,34,36,37,38,40);2-5,7-11H,1H2,(H,12,13)/t20-,21-,22+,23-;2-,3-,4+,5-/m11/s1
Formel vægt 860.3