missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-methylethylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-7031446
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
17797-11-4
C9H12ClN
169.65
MFCD08669636
JCUUNSQUTQELEZ-UHFFFAOYSA-N
2-4-chlorophenyl propan-2-amine, 1-4-chlorophenyl-1-methylethylamine, 2-4-chlorophenyl prop-2-ylamine, 4-chloro-alpha,alpha-dimethylbenzylamine, 4-chlorocumylamine, alpha-amino-4-chlorocumene, 2-4-chlorophenyl-2-propanamine, 2-?4-chlorophenyl?propan-2-amine, benzenemethanamine,4-chloro-, a,, a-dimethyl, benzenemethanamine,4-chloro-a,a-dimethyl
11030256
2-(4-chlorophenyl)propan-2-amine
CC(C)(N)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
17797-11-4
C9H12ClN
169.65
MFCD08669636
JCUUNSQUTQELEZ-UHFFFAOYSA-N
2-4-chlorophenyl propan-2-amine, 1-4-chlorophenyl-1-methylethylamine, 2-4-chlorophenyl prop-2-ylamine, 4-chloro-alpha,alpha-dimethylbenzylamine, 4-chlorocumylamine, alpha-amino-4-chlorocumene, 2-4-chlorophenyl-2-propanamine, 2-?4-chlorophenyl?propan-2-amine, benzenemethanamine,4-chloro-, a,, a-dimethyl, benzenemethanamine,4-chloro-a,a-dimethyl
11030256
2-(4-chlorophenyl)propan-2-amine
CC(C)(N)C1=CC=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 43°C to 45°C (0.3 mmHg)
Odor Amine-like
Quantity 250 mg
UN Number UN2735
Formula Weight 169.66
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-1-methylethylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'