Få mere at vide
1-(4-Chloro-Benzhydryl)-4-Nitroso-Piperazine, TRC
Artikelnummer. 30179797
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179797 100 mg
30180241 25 mg
30180022 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179797 Leverandør Toronto Research Chemicals Leverandørnr. TRCC120540100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2005-04-1
Molekylær formel C17 H18 Cl N3 O
Molekylvægt (g/mol) 315.8
Synonym Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-nitroso-, Piperazine, 1-(p-chloro-α-phenylbenzyl)-4-nitroso- (7CI,8CI), 1-[(4-Chlorophenyl)phenylmethyl]-4-nitrosopiperazine, 1-(4-Chloro-benzhydryl)-4-nitroso-piperazine, 1-[(4-Chlorophenyl)Phenylmethyl]-4-Nitrosopiperazine, N-Nitroso Hydroxyzine Impurity;
IUPAC navn 1-[(4-chlorophenyl)-phenylmethyl]-4-nitrosopiperazine
SMIL Clc1Ccc(Cc1)C(N2Ccn(Cc2)N=O)C3Ccccc3
Tekniske data
Kemisk navn eller materiale 1-(4-Chloro-Benzhydryl)-4-Nitroso-Piperazine
Analyt- eller komponentnavne 1-(4-Chloro-Benzhydryl)-4-Nitroso-Piperazine
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C17H18ClN3O/c18-16-8-6-15(7-9-16)17(14-4-2-1-3-5-14)20-10-12-21(19-22)13-11-20/h1-9,17H,10-13H2
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 100 mg
Formel vægt 315.1138