Få mere at vide
1-(4-Chloro-3-trifluoromethylphenyl)-3-(4-hydroxyphenyl)urea, TRC
Artikelnummer. 30362982
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362982 10 mg
30358046 100 mg
30362140 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362982 Leverandør Toronto Research Chemicals Leverandørnr. TRCC59619510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1129683-83-5
Molekylær formel C14H10ClF3N2O2
Molekylvægt (g/mol) 330.69
Synonym N-[4-Chloro-3-(trifluoromethyl)phenyl]-N'-(4-hydroxyphenyl)urea
IUPAC navn 1-[4-chloro-3-(trifluoromethyl)phenyl]-3-(4-hydroxyphenyl)urea
SMIL Oc1ccc(NC(=O)Nc2ccc(Cl)c(c2)C(F)(F)F)cc1
Tekniske data
Kemisk navn eller materiale 1-(4-Chloro-3-trifluoromethylphenyl)-3-(4-hydroxyphenyl)urea
Mængde 10 mg
InChI formel InChI=1S/C14H10ClF3N2O2/c15-12-6-3-9(7-11(12)14(16,17)18)20-13(22)19-8-1-4-10(21)5-2-8/h1-7,21H,(H2,19,20,22)