missing translation for 'onlineSavingsMsg'
Learn More

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-fluorophenyl)urea, 97%, Alfa Aesar™

Brand:  Alfa Aesar H65673.MD

Additional Details : Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15469826

  • 458.56 DKK / 250mg


Estimated Shipment date
from Supplier 15-01-2019
Add to basket

Specifications

Chemical Identifiers

903238-33-5
332.68
UVICLCVUZCQSQM-UHFFFAOYSA-N
16378533
C14H9ClF4N2O
MFCD07060646
1-4-chloro-3-trifluoromethyl phenyl-3-2-fluorophenyl urea, 4-chloro-3-trifluoromethyl phenyl amino-n-2-fluorophenyl carboxamide

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-fluorophenyl)urea
903238-33-5
MFCD07060646
UVICLCVUZCQSQM-UHFFFAOYSA-N
16378533
Solid
250mg
C14H9ClF4N2O
1-4-chloro-3-trifluoromethyl phenyl-3-2-fluorophenyl urea, 4-chloro-3-trifluoromethyl phenyl amino-n-2-fluorophenyl carboxamide
332.68
332.68
97%
Videos
SDS