missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)azepane, 96%

Brand:  Alfa Aesar™ H63929.06

Additional Details : CAS Number : 40832-89-1 Weight : 0.00500kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15490899

  • 2669.95 DKK / 5g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

40832-89-1
1-(4-Chloro-2-nitrophenyl)hexamethyleneimine

Specifications

1-(4-Chloro-2-nitrophenyl)azepane
40832-89-1
219458
254.71
96%
5g
UN3259
1-(4-Chloro-2-nitrophenyl)hexamethyleneimine
Solid
Videos
SDS Documents