Få mere at vide
1-(4-Chloro-2-hydroxyphenyl)-1,2-dihydro-3H-pyrazol-3-one, TRC
Artikelnummer. 30364675
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364675 50 mg
30356356 250 mg
30364110 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30364675 Leverandør Toronto Research Chemicals Leverandørnr. TRCC42399550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 512165-97-8
Molekylær formel C9 H7 Cl N2 O2
Molekylvægt (g/mol) 210.617
Synonym 3H-Pyrazol-3-one, 1-(4-chloro-2-hydroxyphenyl)-1,2-dihydro-, BF 500-8, 500-M85, M500F085
IUPAC navn 1-(4-chloro-2-hydroxyphenyl)pyrazol-3-ol
SMIL Oc1ccn(n1)c2ccc(Cl)cc2O
Tekniske data
Kemisk navn eller materiale 1-(4-Chloro-2-hydroxyphenyl)-3-hydroxypyrazole
Mængde 50 mg
InChI formel InChI=1S/C9H7ClN2O2/c10-6-1-2-7(8(13)5-6)12-4-3-9(14)11-12/h1-5,13H,(H,11,14)
Formel vægt 210.0196