Få mere at vide
1-(4-Carboxyphenyl)-3-methyl-5-pyrazolone, TRC
Artikelnummer. 30340080
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30340080 50 mg
30334556 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30340080 Leverandør Toronto Research Chemicals Leverandørnr. TRCB49604350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(4-Carboxyphenyl)-3-methyl-5-pyrazolone
CAS 60875-16-3
Molekylær formel C11H10N2O3
Mængde 50 mg
Synonym 4-(4,5-Dihydro-3-methyl-5-oxo-1H-pyrazol-1-yl)benzoic Acid, 1-(4-Carboxyphenyl)-3-methyl-2-pyrazolin-5-one, 1-(4-Carboxyphenyl)-3-methyl-2-pyrazolinone, 1-(p-Carboxyphenyl)-3-methyl-5-pyrazolone, 4-(3-Methyl-5-oxo-2-pyrazolin-1-yl)benzoic Acid, 4-(3-Methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic Acid, 4-(3-Methyl-5-oxopyrazol-1-yl)benzoic Acid
InChI formel InChI=1S/C11H10N2O3/c1-7-6-10(14)13(12-7)9-4-2-8(3-5-9)11(15)16/h2-5H,6H2,1H3,(H,15,16)
SMIL Cc1cc(O)n(n1)c2ccc(cc2)C(=O)O
IUPAC navn 4-(3-methyl-5-oxo-4H-pyrazol-1-yl)benzoic acid
Molekylvægt (g/mol) 218.21
Formel vægt 218.0691