Få mere at vide
1,4-Butanediol Terephthalate, TRC
Artikelnummer. 30344642
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30344642 100 mg
30346651 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30344642 Leverandør Toronto Research Chemicals Leverandørnr. TRCB692670100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Butanediol Terephthalate
CAS 29278-69-1
Molekylær formel C20H18O8
Mængde 100 mg
Synonym 4,4'-((Butane-1,4-diylbis(oxy))bis(carbonyl))dibenzoic Acid, 1,4-Benzenedicarboxylic Acid 1,4-Butanediyl Ester, 1,4-Butanediol bis(Hydrogen Terephthalate), Terephthalic Acid Tetramethylene Ester
InChI formel InChI=1S/C20H18O8/c21-17(22)13-3-7-15(8-4-13)19(25)27-11-1-2-12-28-20(26)16-9-5-14(6-10-16)18(23)24/h3-10H,1-2,11-12H2,(H,21,22)(H,23,24)
SMIL OC(=O)c1ccc(cc1)C(=O)OCCCCOC(=O)c2ccc(cc2)C(=O)O
IUPAC navn 4-[4-(4-carboxybenzoyl)oxybutoxycarbonyl]benzoic acid
Molekylvægt (g/mol) 386.35
Formel vægt 386.1002