Få mere at vide
1,4-Butanediol Dimethylacrylate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1,4-Butanediol Dimethacrylate |
| CAS | 2082-81-7 |
| Molekylær formel | C12 H18 O4 |
| Mængde | 2.5 g |
| Synonym | 2-Propenoic acid, 2-methyl-, 1,4-butanediyl ester (9CI), Methacrylic acid, tetramethylene ester (6CI,7CI,8CI), 1,4-Butanediol, dimethacrylate (8CI), 1,4-Butylene dimethacrylate, 1,4-Butylene glycol dimethacrylate, 1,4-Tetramethylene dimethacrylate, BDDMA, BDMA, Butanediol dimethacrylate, DMM 4, Light Ester 1.4BG, Mhoromer MFM 405, NK Ester BD, Oligotetramethylene glycol dimethacrylate, SR 214, SR 214B, SR 214E, Sartomer 214, Tetramethylene dimethacrylate, Tetramethylene glycol dimethacrylate, Tetramethylene methacrylate, Visiomer 1,4-BDDMA, n-Butane-1,4-diyl dimethacrylate, 1,4-Butanediol Dimethacrylate, 2-Propenoic acid, 2-methyl-, 1,1'-(1,4-butanediyl) ester |
| InChI formel | InChI=1S/C12H18O4/c1-9(2)11(13)15-7-5-6-8-16-12(14)10(3)4/h1,3,5-8H2,2,4H3 |
| SMIL | CC(=C)C(=O)OCCCCOC(=O)C(=C)C |
| IUPAC navn | 4-(2-methylprop-2-enoyloxy)butyl 2-methylprop-2-enoate |
| Molekylvægt (g/mol) | 226.2689 |
| Formel vægt | 226.1205 |