Få mere at vide
1,4-Butanediol Dimethylacrylate, TRC
Artikelnummer. 30179827
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179827 500 mg
30179931 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179827 Leverandør Toronto Research Chemicals Leverandørnr. TRCB710033500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2082-81-7
Molekylær formel C12 H18 O4
Molekylvægt (g/mol) 226.2689
Synonym 2-Propenoic acid, 2-methyl-, 1,4-butanediyl ester (9CI), Methacrylic acid, tetramethylene ester (6CI,7CI,8CI), 1,4-Butanediol, dimethacrylate (8CI), 1,4-Butylene dimethacrylate, 1,4-Butylene glycol dimethacrylate, 1,4-Tetramethylene dimethacrylate, BDDMA, BDMA, Butanediol dimethacrylate, DMM 4, Light Ester 1.4BG, Mhoromer MFM 405, NK Ester BD, Oligotetramethylene glycol dimethacrylate, SR 214, SR 214B, SR 214E, Sartomer 214, Tetramethylene dimethacrylate, Tetramethylene glycol dimethacrylate, Tetramethylene methacrylate, Visiomer 1,4-BDDMA, n-Butane-1,4-diyl dimethacrylate, 1,4-Butanediol Dimethacrylate, 2-Propenoic acid, 2-methyl-, 1,1'-(1,4-butanediyl) ester
IUPAC navn 4-(2-methylprop-2-enoyloxy)butyl 2-methylprop-2-enoate
SMIL CC(=C)C(=O)OCCCCOC(=O)C(=C)C
Tekniske data
Kemisk navn eller materiale 1,4-Butanediol Dimethacrylate
Mængde 500 mg
InChI formel InChI=1S/C12H18O4/c1-9(2)11(13)15-7-5-6-8-16-12(14)10(3)4/h1,3,5-8H2,2,4H3
Formel vægt 226.1205