Få mere at vide
1,4-Butanediol-d10, CDN
Artikelnummer. 30322115
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322115 0.5 g 0.5g
30316284 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322115 Leverandør CDN Leverandørnr. CDND05650.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Butanediol-D10
CAS 71760-76-4
Molekylær formel C4 D10 O2
Mængde 0.5 g
Synonym 1,4-Butane-1,1,2,2,3,3,4,4-d8-diol-d2 (9 CI), 1,4-Butanediol-d10, 1,4-Butanediol-D10
InChI formel InChI=1 S/C4H10O2/c5-3-1-2-4-6/h5-6 H,1-4H2/i1D2,2D2,3D2,4D2,5 D,6 D
SMIL [2 H]OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O[2 H]
IUPAC navn 1,1,2,2,3,3,4,4-octadeuterio-1,4-dideuteriooxybutane
Molekylvægt (g/mol) 100.1826
Formel vægt 100.1308 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity