Få mere at vide
1,4-Butane-1,1,4,4-d4-diol, CDN
Artikelnummer. 30327337
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327337 0.25 g
30316597 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327337 Leverandør CDN Leverandørnr. CDND65030.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Butanediol-1,1,4,4-D4
CAS 36684-44-3
Molekylær formel C4 D4 H6 O2
Mængde 0.25 g
Synonym 1,4-Butane-1,1,4,4-d4-diol (9 CI), 1,1,4,4-Tetradeutero-1,4-butanediol, 1,4-Butanediol-1,1,4,4-d4, 1,4-Butanediol-1,1,4,4-D4, 1,4-Butanediol-d4
InChI formel InChI=1 S/C4H10O2/c5-3-1-2-4-6/h5-6 H,1-4H2/i3D2,4D2
SMIL [2 H]C([2 H])(O)CCC([2 H])([2 H])O
IUPAC navn 1,1,4,4-tetradeuteriobutane-1,4-diol
Molekylvægt (g/mol) 94.1457
Formel vægt 94.0932 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity