Få mere at vide
1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine, TRC
Artikelnummer. 30362241
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30362241 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362241 Leverandør Toronto Research Chemicals Leverandørnr. TRCD416027100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine
CAS 709608-92-4
Molekylær formel C4 2H8 H4 N2
Mængde 100 mg
Synonym 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine (9CI), 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine, 1,4-Butane-d8-diamine, 1,4-Butanediamine-(1,1,2,2,3,3,4,4-D8), 1,1,2,2,3,3,4,4-Octadeuteriobutane-1,4-diamine
InChI formel InChI=1S/C4H12N2/c5-3-1-2-4-6/h1-6H2/i1D2,2D2,3D2,4D2
SMIL [2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C([2H])([2H])N
IUPAC navn 1,1,2,2,3,3,4,4-octadeuteriobutane-1,4-diamine
Molekylvægt (g/mol) 96.2
Formel vægt 96.1503