missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Bromophenyl)naphthalene, 98%

Brand:  Alfa Aesar™ H64193.06

Additional Details : CAS Number : 204530-94-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15401299

  • 2403.83 DKK / 5g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Bromophenyl)naphthalene
283.17
3256523
204530-94-9
MFCD07369916

Specifications

1-(4-Bromophenyl)naphthalene
5g
98%
MFCD07369916
204530-94-9
Solid
283.17
3256523
Videos
SDS