missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromobenzyl)piperazine, 97%, Thermo Scientific™

Catalog Number 15481298
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
91345-62-9
C11H15BrN2
255.159
MFCD02177416
MAHWBNAOEVAPJF-UHFFFAOYSA-N
1-4-bromobenzyl piperazine, 1-4-bromophenyl methyl piperazine, 1-4-bromo-benzyl-piperazine, piperazine, 1-4-bromophenyl methyl, cbmicro_035222, acmc-20apb8, cambridge id 5862395, 1-4-bromobenzyl-piperazine, 4-bromophenyl methyl piperazine
876494
1-[(4-bromophenyl)methyl]piperazine
C1CN(CCN1)CC2=CC=C(C=C2)Br
This item is not returnable. View return policy
91345-62-9
C11H15BrN2
255.159
MFCD02177416
MAHWBNAOEVAPJF-UHFFFAOYSA-N
1-4-bromobenzyl piperazine, 1-4-bromophenyl methyl piperazine, 1-4-bromo-benzyl-piperazine, piperazine, 1-4-bromophenyl methyl, cbmicro_035222, acmc-20apb8, cambridge id 5862395, 1-4-bromobenzyl-piperazine, 4-bromophenyl methyl piperazine
876494
1-[(4-bromophenyl)methyl]piperazine
C1CN(CCN1)CC2=CC=C(C=C2)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 56°C to 62°C
Color Brown to White
Boiling Point 112°C to 116°C (0.3 mmHg)
Quantity 250 mg
UN Number UN3259
Formula Weight 255.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Bromobenzyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'