missing translation for 'onlineSavingsMsg'
Learn More
1-(4-Bromo-2-thienyl)ethan-1-one, Tech., Thermo Scientific™ Product Code.: 10384382

1-(4-Bromo-2-thienyl)ethan-1-one, Tech., Thermo Scientific™

Product Code. p-2694288
1 g, Amber Glass Bottle, 1g
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10384382

Brand: Thermo Scientific Maybridge KM10200DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7209-11-2
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.07
MDL Number MFCD01566552
InChI Key HXVLWNKFMNRJED-UHFFFAOYSA-N
Synonym 1-4-bromothiophen-2-yl ethanone, 2-acetyl-4-bromothiophene, 1-4-bromo-2-thienyl ethan-1-one, 1-4-bromo-2-thienyl-1-ethanone, 1-4-bromothiophen-2-yl ethan-1-one, 4-bromo-2-acetylthiophene, 1-4-bromo-2-thienyl ethanone, ethanone,1-4-bromo-2-thienyl, ethanone, 1-4-bromo-2-thienyl, 4-bromo-2-acetyl-thiopheneShow MoreShow Less
PubChem CID 2735237

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.07
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-(4-Bromo-2-thienyl)ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(4-Bromo-2-thienyl)ethan-1-one, Tech., Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.