Få mere at vide
1,4-Bis(tert-boc)piperazine-d8, TRC
Artikelnummer. 30337971
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30337971 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337971 Leverandør Toronto Research Chemicals Leverandørnr. TRCB41415725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Bis(tert-boc)piperazine-d8
CAS 1246816-49-8
Molekylær formel C14H18D8N2O4
Mængde 25 mg
Synonym 1,4-Bis(tert-butoxycarbonyl)piperazine-d8, 1,4-(Piperazine-d8)dicarboxylic Acid 1,4-Bis(1,1-dimethylethyl)ester
InChI formel InChI=1S/C14H26N2O4/c1-13(2,3)19-11(17)15-7-9-16(10-8-15)12(18)20-14(4,5)6/h7-10H2,1-6H3/i7D2,8D2,9D2,10D2
SMIL [2H]C1([2H])N(C(=O)OC(C)(C)C)C([2H])([2H])C([2H])([2H])N(C(=O)OC(C)(C)C)C1([2H])[2H]
IUPAC navn ditert-butyl 2,2,3,3,5,5,6,6-octadeuteriopiperazine-1,4-dicarboxylate
Molekylvægt (g/mol) 294.42
Formel vægt 294.2395