Learn More
1,4-Bis(diphenylphosphino)butane, 98%, ACROS Organics™
Brand: Acros Organics 296460050
Additional Details : CAS Number : 7688-25-7 Weight : 0.05500kg
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
7688-25-7 | |
426.47 | |
BCJVBDBJSMFBRW-UHFFFAOYSA-N | |
82124 | |
C1=CC=C(C=C1)P(CCCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4 |
C28H28P2 | |
MFCD00003051 | |
1,4-bis diphenylphosphino butane, dppb, phosphine, 1,4-butanediylbis diphenyl, 4-diphenylphosphanyl butyl diphenylphosphane, unii-35hp6ltd2d, tetramethylenebis diphenylphosphine, 35hp6ltd2d, 4-diphenylphosphanylbutyl diphenyl phosphane, 1,4-bis-diphenylphosphino butane | |
4-diphenylphosphanylbutyl(diphenyl)phosphane |
Specifications
1, 4-Bis(diphenylphosphino)butane | |
Glass bottle | |
130.0°C to 137.0°C | |
98% | |
97.5 | |
97.5% min. (HPLC) | |
MFCD00003051 | |
1,4-bis diphenylphosphino butane, dppb, phosphine, 1,4-butanediylbis diphenyl, 4-diphenylphosphanyl butyl diphenylphosphane, unii-35hp6ltd2d, tetramethylenebis diphenylphosphine, 35hp6ltd2d, 4-diphenylphosphanylbutyl diphenyl phosphane, 1,4-bis-diphenylphosphino butane | |
C1=CC=C(C=C1)P(CCCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4 | |
426.47 | |
426.47 | |
98% |
Authentic | |
White | |
5g | |
7688-25-7 | |
100.0 | |
C28H28P2 | |
15, 1254 | |
BCJVBDBJSMFBRW-UHFFFAOYSA-N | |
4-diphenylphosphanylbutyl(diphenyl)phosphane | |
82124 | |
Crystalline Powder |