Få mere at vide
1,4-Bis[3,4-dihydro-2(1H)-quinolinon-7-oxy]butane, TRC
Artikelnummer. 30136012
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136012 250 mg
17959004 100 mg
17909004 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136012 Leverandør Toronto Research Chemicals Leverandørnr. TRCB426500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 882880-12-8
Molekylær formel C22 H24 N2 O4
Molekylvægt (g/mol) 380.44
Synonym 2(1H)-Quinolinone, 7,7'-[1,4-butanediylbis(oxy)]bis[3,4-dihydro-, ,7'-[1,4-Butanediylbis(oxy)]bis[3,4-dihydro-2(1H)-quinolinone], 1,4-Bis(3,4-dihydro-2(1H)-quinolinone-7-oxy)butane
IUPAC navn 7-[4-[(2-oxo-3,4-dihydro-1H-quinolin-7-yl)oxy]butoxy]-3,4-dihydro-1H-quinolin-2-one
SMIL O=C1CCc2ccc(OCCCCOc3ccc4CCC(=O)Nc4c3)cc2N1
Tekniske data
Kemisk navn eller materiale 1,4-bis[3,4-dihydro-2(1H)-quinolinon-7-oxy]butane
Mængde 250 mg
InChI formel InChI=1S/C22H24N2O4/c25-21-9-5-15-3-7-17(13-19(15)23-21)27-11-1-2-12-28-18-8-4-16-6-10-22(26)24-20(16)14-18/h3-4,7-8,13-14H,1-2,5-6,9-12H2,(H,23,25)(H,24,26)
Formel vægt 380.1736