Få mere at vide
1,4-Bis[3,4-dihydro-2(1H)-quinolinon-7-oxy]butane-d8, TRC
Artikelnummer. 30332107
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30332107 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30332107 Leverandør Toronto Research Chemicals Leverandørnr. TRCB42650210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Bis[3,4-dihydro-2(1H)-quinolinon-7-oxy]butane-d8
CAS 1346599-01-6
Molekylær formel C22H16D8N2O4
Mængde 10 mg
Synonym 7,7'-[1,4-(Butane-d8)diylbis(oxy)]bis[3,4-dihydro-2(1H)-quinolinone
InChI formel InChI=1S/C22H24N2O4/c25-21-9-5-15-3-7-17(13-19(15)23-21)27-11-1-2-12-28-18-8-4-16-6-10-22(26)24-20(16)14-18/h3-4,7-8,13-14H,1-2,5-6,9-12H2,(H,23,25)(H,24,26)/i1D2,2D2,11D2,12D2
SMIL [2H]C([2H])(Oc1ccc2CCC(=O)Nc2c1)C([2H])([2H])C([2H])([2H])C([2H])([2H])Oc3ccc4CCC(=O)Nc4c3
IUPAC navn 7-[1,1,2,2,3,3,4,4-octadeuterio-4-[(2-oxo-3,4-dihydro-1H-quinolin-7-yl)oxy]butoxy]-3,4-dihydro-1H-quinolin-2-one
Molekylvægt (g/mol) 388.49
Formel vægt 388.2238