Få mere at vide
1,4-Bis[(1S)-1-(4-methoxyphenyl)ethyl]-2,5-piperazinedione-d4, TRC
Artikelnummer. 30337208
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30337208 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337208 Leverandør Toronto Research Chemicals Leverandørnr. TRCB48602750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Bis[(1S)-1-(4-methoxyphenyl)ethyl]-2,5-piperazinedione-d4
CAS 1246496-54-7
Molekylær formel C8 H17 N O5
Mængde 50 mg
InChI formel InChI=1S/C8H17NO5/c9-6-4(11)1-2-14-8(6)7(13)5(12)3-10/h4-8,10-13H,1-3,9H2/t4?,5-,6?,7?,8?/m1/s1
SMIL NC1C(O)CCOC1C(O)[C@H](O)CO
IUPAC navn (2R)-1-(3-amino-4-hydroxyoxan-2-yl)propane-1,2,3-triol
Molekylvægt (g/mol) 207.224
Formel vægt 207.111