Få mere at vide
1,4’-Bipiperidine-2,2,3,3,4,4,5,5,6,6-d10, TRC
Artikelnummer. 30345702
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30345702 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345702 Leverandør Toronto Research Chemicals Leverandørnr. TRCB398962100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4'-Bipiperidine-2,2,3,3,4,4,5,5,6,6-d10
CAS 718613-20-8
Molekylær formel C10H10D10N2
Mængde 100 mg
InChI formel InChI=1S/C10H20N2/c1-2-8-12(9-3-1)10-4-6-11-7-5-10/h10-11H,1-9H2/i1D2,2D2,3D2,8D2,9D2
SMIL [2H]C1([2H])N(C2CCNCC2)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 2,2,3,3,4,4,5,5,6,6-decadeuterio-1-piperidin-4-ylpiperidine
Molekylvægt (g/mol) 178.34
Formel vægt 178.2254