Få mere at vide
[1,4'-Bipiperidine]-1'-carbonyl-d10 Chloride Hydrochloride, TRC
Artikelnummer. 30339877
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30339877 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339877 Leverandør Toronto Research Chemicals Leverandørnr. TRCB3989675MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale [1,4'-Bipiperidine]-1'-carbonyl-d10 Chloride Hydrochloride
CAS 718613-22-0
Molekylær formel C11H10D10Cl2N2O
Mængde 5 mg
InChI formel InChI=1S/C11H19ClN2O.ClH/c12-11(15)14-8-4-10(5-9-14)13-6-2-1-3-7-13;/h10H,1-9H2;1H/i1D2,2D2,3D2,6D2,7D2;
SMIL Cl.[2H]C1([2H])N(C2CCN(CC2)C(=O)Cl)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 4-(2,2,3,3,4,4,5,5,6,6-decadeuteriopiperidin-1-yl)piperidine-1-carbonyl chloride;hydrochloride
Molekylvægt (g/mol) 277.26
Formel vægt 276.158